Alfa Aesar™ Europium(III) oxide, nanopowder, 99.9% (REO)

Manufacturer: Alfa Aesar™ 4466706

Quantity 5g

 View more versions of this product

Catalog No. AA4466706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1308-96-9
    Molecular Formula Eu2O3
    Formula Weight 351.92
    MDL Number MFCD00010997
    Merck Index 14,3904
    Synonym Europium sesquioxide
    Chemical Name or Material Europium (III) oxide

    Specifications

    Formula Weight 351.92
    Molecular Formula Eu2O3
    TSCA TSCA
    Sensitivity Hygroscopic
    MDL Number MFCD00010997
    CAS 1308-96-9
    Merck Index 14,3904
    Synonym Europium sesquioxide
    Particle Size 45 to 60nm APS
    Surface Area 14 to 18m2/g
    Melting Point 2291°C
    Quantity 5g
    Physical Form Nanopowder
    Chemical Name or Material Europium (III) oxide
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit