Alfa Aesar™ Europium(III) oxide, REacton™, 99.996% (REO)

Manufacturer: Alfa Aesar™ 4434714

Quantity 25g

 View more versions of this product

Catalog No. AA4434714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1308-96-9
    Molecular Formula Eu2O3
    Formula Weight 351.92
    MDL Number MFCD00010997
    Merck Index 14,3904
    Chemical Name or Material Europium (III) oxide
    Grade Reacton™

    Specifications

    Formula Weight 351.92
    Quantity 25g
    TSCA TSCA
    Molecular Formula Eu2O3
    MDL Number MFCD00010997
    Physical Form Powder
    Sensitivity Hygroscopic
    CAS 1308-96-9
    Merck Index 14,3904
    Melting Point 2291°C
    Chemical Name or Material Europium (III) oxide
    Assay 99.996%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit