Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) sulfate octahydrate, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
10031-55-7
Eu2H16O20S3
736.216
MFCD00149704
PGDXJFDXKJLVTJ-UHFFFAOYSA-H
europium iii sulfate octahydrate,2eu.3so4.8h2o,europium sulfate,europium sulfate,,dieuropium 3+ octahydrate trisulfate,europium 3+ sulfate-water 2/3/8
25021676
europium(3+);trisulfate;octahydrate
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3]
10031-55-7
Eu2H16O20S3
736.216
MFCD00149704
PGDXJFDXKJLVTJ-UHFFFAOYSA-H
europium iii sulfate octahydrate,2eu.3so4.8h2o,europium sulfate,europium sulfate,,dieuropium 3+ octahydrate trisulfate,europium 3+ sulfate-water 2/3/8
25021676
europium(3+);trisulfate;octahydrate
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3]
for ultra trace analysis
Assay Percent Range 99.99%
Linear Formula Eu2(SO4)3·8H2O
Merck Index 10, 3854
Formula Weight 736.22
Physical Form Fine Crystalline Powder
Grade Trace Metal
Density 4.95g/mL
Packaging Glass bottle
Total Trace Metal Impurities 0.01% max.
Color White
Quantity 1g
Chemical Name or Material Europium(III) sulfate octahydrate, 99.99%
Show More Show Less