Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium ingot, 99.9% (REO), Thermo Scientific™

Click to view available options
Quantity:
1 g
5 g
7440-53-1
Eu
151.96
MFCD00010992,MFCD00085323
OGPBJKLSAFTDLK-UHFFFAOYSA-N
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
23981
CHEBI:32999
europium
[Eu]
7440-53-1
Eu
151.96
MFCD00010992,MFCD00085323
OGPBJKLSAFTDLK-UHFFFAOYSA-N
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
23981
CHEBI:32999
europium
[Eu]
  • Packaged under mineral oil
Quantity 5 g
UN Number UN3208
Odor Odorless
Physical Form Ingot, Packaged under mineral oil
Percent Purity 99.9%
Color Gray
Chemical Name or Material Europium ingot
Assay (REO)

GHS H Statement
H250-H261

P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H250-H261-H500

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

EINECSNumber : 231-161-7

TSCA : Yes

storageNote1 :

  • Packaged under mineral oil

Recommended Storage : Ambient temperatures

RUO – Research Use Only