Learn More
Novus Biologicals™ Europium Midi Antibody Labeling Kit
Novus Biologicals™ Europium Midi Antibody Labeling Kit allows antibodies or proteins to quickly and easily conjugated to 200nm Europium chelate microspheres.
Manufacturer:  Novus Biologicals™ 12000120
Catalog No. 12-000-120
Add to cart
Description
Description
Kit Components:
  • 1 Midi vial of 200nm Europium
  • 1 vial of 1x Reaction Buffer A
  • 1 vial of 1x Reaction Buffer B
  • 1 vial of 10x Europium Quencher
  • 1 vial of 1x Resuspension Buffer
Specifications
Specifications
1 Midi vial of 200nm Europium, 1 vial of 1x Reaction Buffer A, 1 vial of 1x Reaction Buffer B, 1 vial of 10x Europium Quencher, 1 vial of 1x Resuspension Buffer
1 reaction
Store at -20° C.
Europium Midi Antibody Labeling Kit
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For research use only.