Alfa Aesar™ Europium tris(d,d-dicampholylmethanate), 97%

Manufacturer: Alfa Aesar™ A11676ME

Quantity 500mg

 View more versions of this product

Catalog No. AAA11676ME

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 52351-64-1
    Molecular Formula C63H105EuO6
    Formula Weight 1110.49
    MDL Number MFCD00135060
    Synonym Eu-dcm; Tris(d,d-dicampholylmethanato)europium
    Chemical Name or Material Europium tris (d,d-dicampholylmethanate)

    Specifications

    Formula Weight 1110.49
    CAS 52351-64-1
    Melting Point ≈220°C
    Quantity 500mg
    Molecular Formula C63H105EuO6
    MDL Number MFCD00135060
    Synonym Eu-dcm; Tris(d,d-dicampholylmethanato)europium
    Chemical Name or Material Europium tris (d,d-dicampholylmethanate)
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit