Learn More
Siemens Healthineers Eurotrol Epoc™ Calibration Verification Fluid
Siemens Healthineers Eurotrol Epoc™ Calibration Verification Fluid is an assayed blood gas, electrolyte and metabolite reference material, to verify precision, accuracy, and linearity of the epoc Blood Analysis System
Manufacturer:  Siemens Healthineers 10736357
Catalog No. 22-347-433
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Developed to verify calibration, operating temperature and other performance characteristics
  • pH, gases, electrolytes and metabolites in one ampule
  • Five (5) levels covering the physiologically relevant range
  • Use only one epoc cartridge for each level
  • Optimized for epoc with physiological target values
Specifications
Specifications
1 to 5
4 x 2.5mL samples