Learn More
Siemens Healthineers Eurotrol GAS-ISE Metabolites
Siemens Healthineers Eurotrol GAS-ISE Metabolites is an assayed blood gas, electrolyte and metabolite reference material to verify the precision and accuracy of the epoc Blood Analysis System
Manufacturer:  Siemens Healthineers 10736355
 View more versions of this product
Catalog No. 22-347-431
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • pH, gases, electrolytes and metabolites in one ampule
  • Three (3) levels covering the physiologically relevant range
  • Use only one epoc cartridge for each control check
  • Optimized for epoc with physiological target values
  • Developed to verify calibration, operating temperature and other performance characteristics
Specifications
Specifications
2
10 x 2.5mL vials