Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVA1/MPZL2 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271553JF549

Catalog No. N271553J549


Add to cart

Description

Description

EVA1/MPZL2 Monoclonal antibody specifically detects EVA1/MPZL2 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

EVA1/MPZL2
OTI2C7
Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
MPZL2
Full length recombinant protein of human EVA1/MPZL2 (NP_658911) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
EVAEpithelial V-like antigen 1EVA1myelin protein zero-like protein 2, myelin protein zero-like 2
Mouse
IgG1
0.1 mL
Primary
10205.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit