Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ever-Lite™ Safety Readers

Pyramex™ Ever-Lite™ Safety Readers with ANSI Z87 safety compliance

Manufacturer:  Pyramex™ SB8620R25

 View more versions of this product

Catalog No. 19-555-678



This item is not returnable. View return policy

Description

Description

  • Soft nose piece for snug fit
  • Semi-frame & slim co-material temple design for lightweight spectacle - 22 gm
  • Optical Class 1 lens offers the clearest and the most panoramic view without obstruction
  • 8 base wraparound lens provides full front & side protection
  • Scratch-resistant, polycarbonate lens provide polycarbonate lens
  • Lens Technology:
    • Visible Light Transmitted - 2%
    • UVA, UVB Blocked - 99%
    • Blue Light Blocked - 99%
    • IR Blocked - 99%
Specifications

Specifications

Polycarbonate
Slim Co-Injected Temples
Gray
+2.50
Polycarbonate
Black
ANSI Z87
Eye Safety
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit