30mL Centrifuge Tubes

Tubes withstand up to 6000 x g

Manufacturer: EVERGREEN SCIENTIFIC INTERNATIONAL INC 2203585030

 View more versions of this product

Catalog No. 05-556-3

  Check Availability
Add to Cart

    Specifications

    Product Type In-Cap Spork
    Capacity 1.01 oz., 30mL
    Capacity (English) 1.01 oz.
    Capacity (Metric) 30mL
    Closure Type In-cap spork only
    Material Polypropylene
    For Use With (Application) General purpose laboratory work, stronger acids (except oxidizing acids) or alkalis, alcohols, organic solvents, chlorinated and aromatic hydrocarbons (except xylene) and ketones
    Includes Spork
    Packaging Bulk
    Quantity 1000/Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit