Learn More
Caplugs™ Evergreen Scientific FPC™ Jumbo™ Fecal Parasite Concentrator Kit
Provides a practical, versatile system to collect, preserve, transport, and process fecal specimens for ova and parasite procedures
Manufacturer:  Caplugs™ 3082000000
Catalog No. 05-558-5
Add to cart
Includes: Preassembled centrifuge tube strainer units; Centrifuge tube caps; Dispensing spoons; 20% Triton™ X-100; Trays; Applicator sticks; Extra 30mL vials; Biohazard bags; Instructions
Description
Description
Simple, clean, efficient device used to concentrate helminth eggs and larvae, protozoan cysts, and to recover coccidian oocysts such as Cryptosporidium parvum and Isospora belli. As a modification of the Ritchie formalin-ether (F-E) method, this device replaces funnel-gauze filtrations with two polypropylene tubes and an interconnected plastic strainer unit.
  • Largest filter matrix of any commercially available fecal concentrating device provides 380 ultra-precision 0.6 x 0.6mm holes
  • Attaches directly to patient sample vials; requires no intermediate preparation prior to concentration, reducing handling and the chance for contamination
  • Because the specimen is completely contained, the odor associated with fecal specimens is virtually eliminated
  • Especially advantageous when the specimen contains only a small number of organisms
  • Manufactured under an exclusive licensing agreement with the National Technical Information Service of the United States government
  • Compatible with virtually all commercially available fecal transport kits
  • One case of kits provides materials for 120 tests
Specifications
Specifications
Preassembled centrifuge tube strainer units, Centrifuge tube caps, dispensing spoons, 20% Triton X-100, trays, applicator sticks, 30mL extra vials, biohazard bags, largest filter matrix with 0.6 x 0.6mm holes and instructions
Fecal Parasite Concentrator kit
Healthcare
120 Case