Learn More
Caplugs™ (Evergreen Scientific) Sera-Separa™ Serum Filters
$537.54 - $578.00
Specifications
Product Type Serum Filter
For Use With (Equipment) EverGreen Serum tubes
Quantity 1000 Pack
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Length (English) Length (Metric) Price Quantity & Availability  
05-551-5
View Documents
Caplugs™ (Evergreen Scientific)
2083193020
0.16 oz. 5mL 2.62 in. 66.5mm
Case of 1000 for $537.54
Add to cart
22-046983
View Documents
Caplugs™ (Evergreen Scientific)
2083084020
0.30 oz. 9mL 4.25 in. 107mm
Case of 1000 for $578.00
Add to cart
Description
Description
  • Flange is inserted into evacuated blood tube and pushed downward
  • Wide body style prevents sample probes from hitting side walls
  • Also for use in removing fibrin and macro particles from serum to prevent line clogging
  • Can be used with Evergreen serum tubes
Specifications
Specifications
Serum Filter
EverGreen Serum tubes
1000 Pack