Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caplugs™ (Evergreen Scientific) Sterile Polypropylene Culture Tubes
Precision molded from premium-grade, chemically inert plastic materials
Supplier: Caplugs™ (Evergreen Scientific) 222239408I
Description
- Gamma radiation sterilized; shipped ready to use
- Tubes are constructed of polypropylene for toughness and chemical resistance
- Polypropylene tubes are almost transparent and withstand centrifugation up to 6000 × g
- Overfitting caps have inner ribs for two-position fits: loose for aerobic culturing and tight for anaerobic culturing and sealing
- Tabs and/or rings allow easy removal and sealing
- Screw caps have built-in sealing rings to provide leakproof closures
- Available with or without caps; packaged in easy-to-open bags of 25 tubes each
- Individual packaging available for 12 × 75mm, 17 × 100mm, and 16 × 125mm sizes (see Cat. Nos. 22-047-511, -514)
Specifications
6000 x g | |
17 mm | |
100 mm | |
Two position cap | |
Sterile |
Cell Culture Tube | |
Dual-Position | |
Polypropylene | |
500/Cs. | |
16 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction