Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI2B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen EVI2B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159952
SDP
View Documents
Novus Biologicals
NBP159952
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

EVI2B Polyclonal specifically detects EVI2B in Human samples. It is validated for Western Blot.
Specifications

Specifications

EVI2B
Polyclonal
Purified
RUO
CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B
EVI2B
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
P34910
2124
Synthetic peptides corresponding to EVI2B(ecotropic viral integration site 2B) The peptide sequence was selected from the middle region of EVI2B. Peptide sequence DICMDNIRENEISTKRTSIISLTPWKPSKSTLLADDLEIKLFESSENIED.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.