Learn More
EVI2B Mouse anti-Human, PE, Clone: MEM-216, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15806
Catalog No. 50-113-7771
Add to cart
Description
Description
CD361, also known as EVI2B (Ecotropic Viral Integration site 2B) or EVDB, is a poorly characterized type I transmembrane protein, expressed from one of three genes embedded in intron 27b of the neurofibromatosis type 1 (NF1) gene. The DNA strand that is transcribed to produce CD361 is the complementary one to the strand encoding NF1. Murine homolog to human CD361 is associated with ecotropic viral insertions, which have been implicated in the expression of murine myeloid leukemias. CD361 has been also reported to be involved in melanocyte and keratinocyte differentiation. However, it is expressed mainly in peripheral blood and bone marrow.
Specifications
Specifications
EVI2B
Monoclonal
PE
EVI2B
Liquid
EVI2B
human CD361
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MEM-216
PBS with 0.1% sodium azide
P34910
D17S376, EVDB, EVI-2B, ecotropic viral integration site 2B protein homolog, protein EVI2B
Mouse
IgG1
100 tests
4° C
Primary
2124