Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI2B Rabbit anti-Human, Polyclonal , Abnova™

Rabbit polyclonal antibody raised against partial recombinant human EVI2B.

Manufacturer:  Abnova Corporation PAB29575

Catalog No. 89-934-113


Add to cart

Description

Description

Sequence: ETPDICMDNIRENEISTKRTSIISLTPWKPSKSTLLADDLEIKLFESSENIEDSNNPKTEKIKDQVNGTSEDSADGSTVGTAVSSSDDADLPPPPPLLDLEGQESNQSDKPTMTIVSPLPNDSTSLPPS
Specifications

Specifications

EVI2B
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) The optimal working dilution should be determined by the end user.
Liquid
EVI2B
Recombinant protein corresponding to amino acids 248-376 of human EVI2B.
Antigen affinity purification
RUO
Primary
2124
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against partial recombinant human EVI2B.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
D17S376/EVDB
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit