Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Evolution™ 300 UV-Vis Spectrophotometer

The Thermo Scientific™ Evolution 300 is ideal for life science, pharmaceutical and QA/QC applications. A wide selection of accessories and a low cost of ownership make it a versatile and affordable addition to the lab.

Manufacturer:  Thermo Scientific™ 840280200

 View more versions of this product

Catalog No. 14-386-407


  Check Availability
Add to cart

Specifications

Description

The Thermo Scientific™ Evolution 300 is ideal for life science, pharmaceutical and QA/QC applications. A wide selection of accessories and a low cost of ownership make it a versatile and affordable addition to the lab. The powerful Xenon lamp-based platform and variable bandwidth make the Evolution 300 ideal for life science, pharmaceutical and QA/QC applications. A selection of VISION software packages expand instrument capability and Smart Accessories make your analyses easier to perform. The walk-up convenience of the Xenon lamp saves valuable time and increases your lab's productivity.

The powerful Xenon lamp-based platform and variable bandwidth make the Evolution 300 ideal for life science, pharmaceutical and QA/QC applications. A selection of VISION software packages expand instrument capability and Smart Accessories make your analyses easier to perform. The walk-up convenience of the Xenon lamp saves valuable time and increases your lab's productivity.

Xenon Lamp

  • Provides excellent performance over the full wavelength range
  • Ready to use without instrument warmup for fast readings
  • Activates only when taking a measurement; extended lamp life lowers cost of instrument ownership
  • 3-year warranty

Key Features

  • 5 variable spectral bandwidth settings for matching customer assays to instrument
  • Acquisition rate: 50 data points per second
  • Longlife Xenon lamp

Optional Smart Accessories For Any Application

  • Full line available for Life Sciences and QA/QC
  • Install or remove without restarting the instrument
  • Built-in software instantly identifies and configures to accommodate accessory
  • Each accessory has a unique serial number that is recorded for comprehensive event logs and audit trail capabilities
  • Automated performance verification with CVC accessory
  • Mercury lamp accessory is available for testing and defining wavelength accuracy

PC Control

  • Enables data collection and distribution to third party applications for advanced processing and archiving
  • Uses powerful VISION software suite to control instrument
  • Collects, stores, recalls and analyzes data

Local and PC Control

Combines keypad and PC control systems for maximum flexibility in labs with a variety of experimental needs

Ordering Information:

All accessories are sold separately. Available software packages include VISIONliteMaterialsCalc (for material analysis), VISIONpro (standard UV-VIS), VISIONlife (Life Science), VISIONsecurity (regulated labs), VISIONlite ColorCalc Basic (XYZ, Lab, CIE), and VISIONlite ColorCalc Advanced (>40DAB/EP color check filters, Pt-Co/Alpha color, Gardner and Iodine color, wine analysis). Other accessories include cell changers, cuvet and test holders, calibration carousels, sippers, fiber optic probes, small volume sampling, and accessories for reflectance and rapid mixing.

Recommended for:

  • General Spectroscopy
  • High-resolution UV-Visible spectrscopy
  • Temperature-based investigations

Life Science

  • Nucleic acid assays
  • Protein concentration assays
  • Thermal denaturation/renaturation
  • Kinetics

QA/QC

  • Quantitative analysis
  • Finished product analysis
  • Measurements in the regulated laboratory

Pharmaceuctical

  • General Research
  • QA/QC
  • Tablet dissolution

Note:

To use the Evolution 300 UV-Vis Spectrophotometer in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Description & Specifications

Specifications

±0.004A at 1A, ±0.004A at 2A, ±0.006A at 3AÅ
±0.0015A (200-800nm), 2.0nm SBW, smoothed
21 CFR Part 11 and audit-proof IQ/OQ/PQ documentation. ISO 9001:2000
21in.
Evolution 300 PC UV-Vis Spectrophotometer
<0.0005Abs/hr, 500 nm, 2.0 nm SBW, 2hr warm-up
15in.
50/60Hz
300 PC w/VisionPro Software
Xenon Flash Lamp
Typical lifetime: >5 years, longer if not using live signal
Warranty period: 3 year source replacement warranty
Modified Ebert

Modified Ebert

Double beam with sample and reference cuvette/accessory positions

Resolution (Toluene in Hexane): Peak/Trough Ratio >2.0

Photometric Accuracy (60mg/L K2Cr2O7): <±0.01 A relative to calibrated sealed standard

Stray Light: <0.13 %T at 198 nm with KCl per EP

220 nm: <0.01%T NaI (behaves identically to KI for USP)

Wavelength Accuracy: ±0.20 nm (546.11 nm Hg emission line), ±0.30 nm 190–900 nm

Wavelength Repeatability: Peak separation of repetitive scanning of Hg line source <0.10 nm

±6 A
Absorbance, % Transmittance, % Reflectance, Concentration
>4A
Absorbance, % Transmittance, % Reflectance, Concentration,
1st-4th Derivative
Variable 0.5; 1.0; 1.5; 2.0; 4.0 nm
At least Windows XP
±0.20nm (546.11nm Hg emission line); ±0.3nm for 190 to 900nm
190 to 1100nm
3800, 2400, 1200, 600, 240, 120, 60, 30, 10, 5, 1 nm/min Intelliscan
22kg
61cm
±0.004A at 1A, ± 0.004A at 2A, ± 0.006A at 3A
Dual-Beam; Quartz Coated
RS-232
53cm
Dual Matched Silicon Photodiodes
100/240V 50/60Hz
38cm
VisionSecurity Software, User Guide and USB Cable
Computer Control
0.5nm

Photometric:

0A: <0.00018 A

1A: <0.00022 A

2A: <0.00050 A

500 nm, 2.0 nm SBW, RMS

Up to 100mm cuvettes

1A: ±0.008 A

2A: ±0.010 A

3A: ±0.018 A

>4A
1A: ±0.0025 A
Peak/Trough >2.0 at 0.5nm SBW
1 to 3800nm/min

198 nm: 2.9 A KCl

220 nm: 4.2 A NaI

340 nm: 4.3 A NaNO2

100/240V
10, 5, 2, 1, 0.5, 0.2, 0.1, 0.05 nm
Standard deviation of 10 measurements <0.05 nm
48.4lb.
24in.
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit