Thermo Scientific™ Evolution™ 300 UV-Vis Spectrophotometer

The Thermo Scientific™ Evolution 300 is ideal for life science, pharmaceutical and QA/QC applications. A wide selection of accessories and a low cost of ownership make it a versatile and affordable addition to the lab.

Manufacturer: Thermo Scientific™ 10300101

 View more versions of this product

Catalog No. 14-386-406

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


Ordering Information:

All accessories are sold separately. Available software packages include VISIONliteMaterialsCalc (for material analysis), VISIONpro (standard UV-VIS), VISIONlife (Life Science), VISIONsecurity (regulated labs), VISIONlite ColorCalc Basic (XYZ, Lab, CIE), and VISIONlite ColorCalc Advanced (>40DAB/EP color check filters, Pt-Co/Alpha color, Gardner and Iodine color, wine analysis). Other accessories include cell changers, cuvet and test holders, calibration carousels, sippers, fiber optic probes, small volume sampling, and accessories for reflectance and rapid mixing.

Recommended for:

  • General Spectroscopy
  • High-resolution UV-Visible spectrscopy
  • Temperature-based investigations

Life Science

  • Nucleic acid assays
  • Protein concentration assays
  • Thermal denaturation/renaturation
  • Kinetics

QA/QC

  • Quantitative analysis
  • Finished product analysis
  • Measurements in the regulated laboratory

Pharmaceuctical

  • General Research
  • QA/QC
  • Tablet dissolution

Note:

To use the Evolution 300 UV-Vis Spectrophotometer in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Specifications

Absorbance, % Transmittance, % Reflectance, Concentration,
1st-4th Derivative
1A: ±0.0025 A
Absorbance, % Transmittance, % Reflectance, Concentration
10, 5, 2, 1, 0.5, 0.2, 0.1, 0.05 nm
Xenon Flash Lamp
Typical lifetime: >5 years, longer if not using live signal
Warrenty period: 3 year source replacement warrenty
Peak/Trough >2.0 at 0.5nm SBW
Standard deviation of 10 measurements <0.05 nm
53 cm
>4A
100/240 V 50/60 Hz
22 kg
190 to 1100nm
24 in.

Photometric:

0A: <0.00018 A

1A: <0.00022 A

2A: <0.00050 A

500 nm, 2.0 nm SBW, RMS

15 in.
At least Windows XP
0.5 nm
300 PC w/VisionSecurity Software
21 in.
38 cm
1 - 3,800 nm/min
48.4 lb.
50/60 Hz

Resolution (Toluene in Hexane): Peak/Trough Ratio >2.0

Photometric Accuracy (60mg/L K2Cr2O7): <±0.01 A relative to calibrated sealed standard

Stray Light: <0.13 %T at 198 nm with KCl per EP

220 nm: <0.01%T NaI (behaves identically to KI for USP)

Wavelength Accuracy: ±0.20 nm (546.11 nm Hg emission line), ±0.30 nm 190–900 nm

Wavelength Repeatability: Peak separation of repetitive scanning of Hg line source <0.10 nm

±6 A

1A: ±0.008 A

2A: ±0.010 A

3A: ±0.018 A

>4 A
3800, 2400, 1200, 600, 240, 120, 60, 30, 10, 5, 1 nm/min Intelliscan
±0.004A at 1A, ± 0.004A at 2A, ± 0.006A at 3A
Modified Ebert
Up to 100 mm cuvettes

Modified Ebert

Double beam with sample and reference cuvette/accessory positions

RS-232
21 CFR Part 11 and audit-proof IQ/OQ/PQ documentation. ISO 9001:2000
Dual-Beam; Quartz Coated
±0.20 nm (546.11 nm Hg emission line); ±0.3 nm for 190 - 900 nm
VisionSecurity Software, User Guide and USB Cable
Evolution 300 Security UV-Vis Spectrophotometer

198 nm: 2.9 A KCl

220 nm: 4.2 A NaI

340 nm: 4.3 A NaNO2

100/240 V
Dual Matched Silicon Photodiodes
±0.0015A (200-800nm), 2.0nm SBW, smoothed
±0.004A at 1A, ±0.004A at 2A, ±0.006A at 3AÅ
Computer Control
Variable 0.5; 1.0; 1.5; 2.0; 4.0 nm
61 cm
<0.0005Abs/hr, 500 nm, 2.0 nm SBW, 2hr warm-up
  • Description
  • Specifications
  • Accessories
  • Description
    Specifications
    Accessories

    Related Products


    Product Content Feedback

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit