Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVORF Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen EVORF
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092971
View Documents
Novus Biologicals
NBP303321100UL
100 μg
Each for $320.71
Add to cart
 
NB092972
View Documents
Novus Biologicals
NBP30332120UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

EVORF Polyclonal antibody specifically detects EVORF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

EVORF
Unconjugated
Rabbit
Human, Mouse, Rat
C16orf80, chromosome 16 open reading frame 80, fSAP23, GTL3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-193 of human EVORF (NP_037374.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
29105
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit