Learn More
EVOS EVOS 50X Oil Objective, achromat, coverslip-corrected
For general applications and image with different magnification
Manufacturer: EVOS AMPFOP050/DEL
Description
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Plan Achromat | |
EVOS™ | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
EVOS® FL Color Imaging System, EVOS® FL Imaging System, EVOS® FL Auto Imaging System | |
0.95 | |
0.19mm |
50X | |
1 each | |
Objective Lens | |
Objective Lens | |
50X |