EVOS™ Digital Color Fluorescence Microscope
All-in-one digital inverted microscope produces extraordinary on-screen images
Manufacturer: EVOS™ AMEFC4302US
 View all options for this product
Catalog No. 12-563-341
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Camera Type Digital color
    Magnification Power 10x, 20x, 40x fluor
    Objective Types 10x, 20x, 40x fluor
    Lighting Type LED
    Optical Design Precision
    Display Type LCD
    Software Type Integrated software
    Unique LED light cube system. Light engine outputs remarkable intensity over a short light path to deliver incomparable excitation. EVOS Light Cubes are easily installed and maintenance-free. Digitally controlled LED light sources allow precise adjustment of illumination levels, dramatically improving control over photobleaching.
    • LED light cubes last >50,000 hours: no need to change bulbs
    • Instant-on; no warm up or cool down required
    • Precision optics for excellent image quality
    • Brightfield and phase contrast
    • Advanced ergonomic design: microscopy made comfortable
    • Small footprint; fits inside cell culture hoods
    • Integrated computer and imaging software
    • High-sensitivity digital camera
    • High-resolution, 15 in. LCD
    • Single switch turns on entire workstation
    • Streamlined software and user interface
  • Description & Specifications
Related Products
Product Content Feedback