Learn More
EVOS™ Olympus 60X Oil Objective, apochromat, coverslip-corrected
For demanding applications, especially capturing color images in white light
Supplier: EVOS™ AMEP4694
Description
- Can be used to view images with different magnification
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Objective Lens | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
Plan Apochromat | |
1 Ea. |
Olympus | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
60X |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.