Learn More
Invitrogen™ EVOS™ Onstage Incubator (OSI-2) Filter Replacement Kit
Description
The EVOS Onstage Incubator (OSI-2) Filter Replacement Kit contains everything needed for the self-installation of new OSI-2 foam filters. These filters are recommended to be replaced every two years.
Specifications
Specifications
| Type | Onstage Accessory |
| Product Line | EVOS |
| Description | EVOS™ OSI-2 Foam Filter Replacements |
| For Use With (Equipment) | EVOS™ Onstage Incubator (OSI-2) |
| Content And Storage | Two foam filters |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.