Learn More
Invitrogen™ EVOS™ Onstage Incubator (OSI-2) Master Plate
Description
The EVOS Onstage Incubator (OSI-2) Master Plate is specifically designed to hold the OSI-2 environmental chamber when used with either the EVOS M5000 Imaging System or EVOS M7000 Imaging System. The OSI-2 master plate is a component of the OSI-2 (link to new PDP page for AMC2000), offered separately in the event the stage plate needs replacing. Only vessel holders designed for the EVOS Onstage Incubator (link to vessel holder selection guide, currently being updated or online selection tool if the latter is available) should be used. This master plate, together with the OSI-2 vessel holder, accommodates various types of cell culture vessels and provides flexibility in your approach to extended time-lapse imaging experiments under precisely controlled environmental conditions.
Specifications
Specifications
| Type | Onstage Accessory |
| Product Line | EVOS |
| Description | EVOS™ OSI-2 Master Plate |
| For Use With (Equipment) | EVOS™ Onstage Incubator (OSI-2) |
| Holds | Onstage incubator compatible vessel holders & vessel |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.