Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVPL Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259098
Description
EVPL Polyclonal specifically detects EVPL in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
EVPL | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
210 KDa Cornified Envelope Precursor Protein, 210 KDa Paraneoplastic Pemphigus Antigen, envoplakin, EVPK, p210 | |
Rabbit | |
Affinity Purified | |
RUO | |
2125 | |
Store at 4C. |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
EVPL | |
Maps to a region between 1275 and 1325 | |
100 μL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only