Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EVX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578385
Description
Positive Control: mouse brain, rat brain Predicted Reactivity: Human (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EVX2, a protein related to the Drosophila even-skipped (eve), is expressed in limbs like the neighbouring Hoxd genes. Evx-2 functions like a Hoxd gene.
Specifications
EVX2 | |
Polyclonal | |
Unconjugated | |
EVX2 | |
eve, even-skipped homeo box homolog 2; even skipped homeotic gene 2; even skipped homeotic gene 2 homolog; even-skipped homeo box 2 (homolog of Drosophila eve); even-skipped homeobox 2; Evx2; Evx-2; Homeobox even-skipped homolog protein 2; LOW QUALITY PROTEIN: homeobox even-skipped homolog protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14029, 288155 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.82 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P49749 | |
EVX2 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse EVX2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction