Learn More
Cole-Parmer Parker Hannifin CV-1-1144 Poppet Check Valve, 1/4" NPT(F) connections
Manufacturer:  Cole-Parmer 0637357
Encompass_Preferred
Valves have body made of PFA and O-ring seals made of Viton. Max pressure is 100 psi; temperature range is 0 to 300°F (–18 to 149°C). Install in any position.
Note: Valves require 5 psi back pressure to achieve a proper seal.
Catalog No. NC1354549