Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EWSR1 Antibody (5H7), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP192686
Description
EWSR1 Monoclonal specifically detects EWSR1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
EWSR1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 | |
Q01844 | |
EWSR1 | |
Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProt# Q01844] | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Equine | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
5H7 | |
Unconjugated | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS | |
Mouse | |
80 kDa | |
0.1 mL | |
Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
2130 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction