Learn More
EWSR1 Mouse anti-Human, Mouse, Rat, Canine, DyLight 550, Clone: 5H7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192686R
Catalog No. NBP192686R
Add to cart
Description
Description
EWSR1 Monoclonal antibody specifically detects EWSR1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
EWSR1
5H7
Western Blot, Immunocytochemistry/Immunofluorescence
bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Canine, Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
EWSR1
Full length recombinant human EWSR1 expressed in and purified from E. coli. [UniProtNo. Q01844]
0.1 mL
Primary
2130