Learn More
DWK Life Sciences Kimble™ Exax Brand Nessler Tall-Form Color Comparison Tubes with standard taper Cap Stoppers Available on GSA/VA Contract for Federal Government customers only.
With standard taper Cap Stoppers
Manufacturer:  DWK Life Sciences 45325A50
 View more versions of this product
Catalog No. 07-059A
Add to cart
Description
Description
With standard taper Cap Stoppers
  • Stopper has shadowless bottom so it need not be removed when making readings
  • The word “shadowless” is printed on side of all caps to distinguish them from regular weighing stoppers
  • 50 to 100mL size designed from ASTM D 1209-00 (Kimble 45325A-50; 45325B-50100)
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Colorless
Standard Taper Stopper
20mm
50mL
White ceramic enamel rings and legend
19/10
7.87 to 8.66 in.
Borosilicate Glass
312mm
NNN-T-175, USP, ASTM E 438, APHA
Used to fill with standard solution colors for visual color comparison with similar tubes containing solution sample
50mL
Graduated Ring Scale