Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare ExcelGel 12.5% Tris-Acetate Gel, 1-well Available on GSA/VA Contract for Federal Government customers only.

Separation Range: 10 to 120 kDa; Separation Type: Denaturing; For Use With: Multiphor II Electrophoresis Unit, ExcelGel SDS BufferStrips

Manufacturer:  GE Healthcare 17600221

Catalog No. 45-002-381


Add to cart

Description

Description

Precast polyacrylamide gels make a wide range of electrophoretic techniques readily available with minimum preparation. The manufacturing quality and reproducibility of these gels translate into highly reliable and reproducible results with minimal training.
  • Perform second-dimension separations using convenient precast gels and 7-, 11-, 18-, or 24-cm IPG strips
  • ExcelGel precast SDS-polyacrylamide gel is available as a 12.5% homogeneous gel
  • Prepare multiple 2-D maps on a single gel using three 7cm or two 11cm IPG strips side-by-side
  • Configure Multiphor II with MultiTemp III Thermostatic Circulator and the programmable EPS 1001 power supply for reproducible running conditions.
Multiphor II Buffer Strip Positioner facilitates placement of ExcelGel Buffer Strips and holds the strips securely in place during the run. CleanGel, Immobiline DryPlate, and DryStrip products are supplied dry and must be rehydrated in the appropriate buffer before use. Prepared swelling solutions are available for the most common procedures. All precast gel recipes are proprietary. However, buffer formulations for a variety of other applications are included in the instructions.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

6/pk
12.5%
10 to 120 kd
1-well
33mm, 6/3 % T/% C
Precast Gel
Multiphor II Electrophoresis Unit, ExcelGel SDS BufferStrips
Tris-Acetate
Denaturing
SDS-PAGE
2 - 8°C
Safety and Handling

Safety and Handling

ShelfLife : 15 Months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit