Learn More
GE Healthcare ExcelGel DNA Analysis Kit Available on GSA/VA Contract for Federal Government customers only.
Designed for use with the Multiphor II Electrophoresis System. GE Healthcare ExcelGel DNA Analysis Kit contains thin polyacrylamide gels and a discontinuous buffer system that offer significantly improved resolution over agarose gels.
Manufacturer:  GE Healthcare 17119807
Catalog No. 45-002-367
Add to cart
Description
Description
DNA Analysis Kit for use especially with the Multiphor II Electrophoresis System
  • Kit includes five ready-to-use gels (12.5% acrylamide) and six pairs of buffer strips
  • Gels and buffers constitute a discontinuous buffer system suitable for most applications
  • Contains 48 sample wells
Usage Notes:
Apply kerosene or silicone oil between the gel and the separation bed of the electrophoresis unit to ensure good thermal contact. In addition, precool (or preheat) the gel before applying the sample.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1 Each
DNA Analysis Kit