Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare ExcelGel 15% Tris-Acetate plus Ethylene Glycol Gel, 25-well Available on GSA/VA Contract for Federal Government customers only.

Separation Range: 3 to 8 kDa; Separation Type: Denaturing; For Use With: Immobiline DryStrip Gels

Manufacturer:  GE Healthcare 80126201

Catalog No. 45-002-431


Add to cart

Description

Description

ExcelGel SDS homogeneous gels have 25 preformed sample wells for sample volumes from 5 to 10µl. They feature a 33mm stacking zone and a 77mm separating zone.
  • Save time with ready-to-use, mechanically stable gels cast on plastic supports
  • Apply samples quickly and easily using preformed sample wells
  • Avoid making running buffer by using precast buffer strips

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

6/pk
Tris-Acetate, plus Ethylene Glycol
ExcelGel precast gels for SDS-PAGE
110 mm
7.5% T/ 3% C
Precast Gel
15%
25-well
Analytical chemistry, Research Horizontal electrophoresis of SDS denatured proteins
110 mm
2 - 8°C
Polyester film
Safety and Handling

Safety and Handling

ShelfLife : 15 Months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit