Learn More
METTLER TOLEDO™ Excellence XPR Analytical Balances
METTLER TOLEDO™ Excellence XPR Analytical Balances provide ultimate accuracy and quality assurance functions that are second to none.
Manufacturer:  METTLER TOLEDO™ 30355415
 View more versions of this product
Catalog No. 01-919-125
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
XPR Analytical Balances, succesor to the XPE Analytical Balances, delivers ultimate weighing performance and a range of intelligent quality assurance functions that fulfill the most stringent requirements.  Depend on XPR to deliver right-first-time results. Patented StaticDetect™ technology, used in combination with the optional ionizing module, offers a complete electrostatic detection and elimination system to ensure accurate weighing results and highest process security.
Optional modules and accessories make it easy to customize your XPR analytical balance to perfectly fit your process needs.
Accurate Results
  • Active Temperature Control system (ATC™)
  • StaticDetect™
  • proFACT Advanced internal adjustment
  • Ionizer module ready
Efficient Operations
  • 7-inch color touchscreen (glove compatible)
  • ErgoClip for direct dosing
  • SmartSens for touch-less door operation
  • SmartTrac dosing guide
  • SmartGrid weighing pan
  • Results protocol
  • Storage of methods
  • Automatic draft shield doors
Quality Assurance
  • MinWeigh warning function
  • Balance ready StatusLight
  • LevelControl: Level warning with graphical levelling guide
  • Tolerance profiles
  • User management
  • Adjustment and routine test history
  • Change history function
Seamless Process
  • LabX ready
  • Calibry ready
  • Multiple ports for easy connectivity and data export: 4 x USB, 1 x LAN
  • Upgradable to automatic powder and / or liquid dosing
Specifications
Specifications
81 g / 220 g
GLP, ISO 9001, UL, CSA
TFT Color Touchscreen
Die-cast aluminum
1.5 s
110/230V 50/60Hz
46 cm
37cm
Analytical Balance
0.01 mg / 0.1 mg
balance, power cable, user manual, production certificate, declaration of conformity, ErgoClip basket, SmartPrep (2), tweezers, spatula (2), brush
53 cm
12.260 kg
USB, LAN, PS/2, RS232 compatible
Analytical Balance XPR205DR