Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Mettler Toledo™ Excellence XSR Precision Balances, Standard Weigh Pan

XSR precision balances offer a winning combination of reliable performance and truly ergonomic operation. Even under the harshest of weighing conditions, you can rely on these robust balances to help you meet your productivity targets.

Manufacturer:  Mettler Toledo™ 30317487

 View more versions of this product

Catalog No. 01-919-054



Description

Description

  • SmartPan weigh pan technology on 0.01g and 0.001g balances
  • 4.3 in. color capacitive touchscreen terminal, glove compatible
  • Storage of weighing methods
  • Integrated results notepad
  • Multiple interfaces for easy connectivity and data export: 4 x USB, 1 LAN, 1 RS232
  • Easy connection of peripheral devices: printers, PC, barcode readers, footswitches and sensors
  • Export of process and results data to XML or Excel
  • Customizable tolerance profiles, with pre-defined quality and weighing settings
  • MinWeigh warning function
  • Balance ready StatusLight™
  • LevelControl: level warning with graphical leveling guide
  • Temperature and time programmable automatic adjustment (proFACT)
  • Password protected configuration
  • Integrated overload protection
  • Chemically resistant materials
  • Designed for easy cleaning
Specifications

Specifications

10 kg
FDA 21 CFRpart 11
XSR10001S
USB, LAN, RS232 compatible
0.1 g
Color Graphic Display
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit