Learn More
Excelta™ Excelta™ Extreme Precision Stainless Steel Tweezers with Straight Soft Replaceable Acetal Tips
Acetal tips offers resistance to chemicals and solvents
Supplier: Excelta™ 169BRTW
Description
- For cleanroom use
- Extreme Precision: Four star
- Overall length: 125mm (5 in.)
Specifications
Straight | |
125 mm | |
1 mm |