Learn More
Excelta™ Excelta™ Fine Precision (Two Star) Stainless Steel Dressing Forceps
Features serrated grips for better control
Manufacturer:  Excelta™ 23DSSE
 View more versions of this product
Catalog No. 18-100-915
Add to cart
Description
Description
  • Made of anti magnetic Stainless steel
  • Straight serrated square point
Specifications
Specifications
Straight Serrated, Square
Stainless steel
Forceps, Dressing
1.7 mm
0.37 in.
4.5 in.
115 mm
0.07 in.
Fine
9.28 mm