Learn More
Excelta™ Five Star Porta-Wand Elite Wafer Pickup System
Vacuum handling tools, kits and accessories
Manufacturer:  Excelta™ PV500W12
 View more versions of this product
Catalog No. 17-468-521
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Vacuum pick-up hand-piece
  • Stainless steel wafer wand
  • Lightweight
  • With push-button
  • Can be used with press-fit tips, adapters and vacuum hoses
  • For 0.13 in. (3.2mm) I.D. hose
Vacuum tool kit contents:
  • Vacuum pick-up hand-piece
  • Detachable 9.6V NiMH rechargeable battery pack
  • In-stand charger
  • Rear exhaust filter
  • Power-on indicator
  • Push button with on/off and lock-to-on
  • Intake air filter
  • Choice of wafer tip
Specifications
Specifications
Ideal for handling wafers, disk media, flat panels, any item that has a flat hard surface
Press fit tips, adapters and vacuum hoses
Vacuum pickup hand-piece, detachable 9.6V NiMH rechargeable battery pack, In stand charger, rear exhaust filter, power on indicator, push button with on/off and lock to on, intake air filter, 12 in. wafer tip
Vacuum Tool Kit