Excelta™ Five Star Porta-Wand Elite Wafer Pickup System
Vacuum handling tools, kits and accessories
Manufacturer: Excelta CorporationSupplier Diversity Partner PV500W6
 View more versions of this product
Catalog No. 17-468-519
  Check Availability
Add to Cart
    Specifications
    Includes Vacuum pickup hand-piece, detachable 9.6V NiMH rechargeable battery pack, In stand charger, rear exhaust filter, power on indicator, push button with on/off and lock to on, intake air filter, 6 in. wafer tip
    Product Type Vacuum Tool Kit
    For Use With (Application) Ideal for handling wafers, disk media, flat panels, any item that has a flat hard surface
    For Use With (Equipment) Press fit tips, adapters and vacuum hoses
  • Description & Specifications
Related Products
Product Content Feedback