Learn More
Excelta™ Five Star Porta-Wand Elite Wafer Pickup System
Vacuum handling tools, kits and accessories
Manufacturer:  Excelta CorporationSupplier Diversity Partner PV500W8
 View more versions of this product
Catalog No. 17-468-520
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Ideal for handling wafers, disk media, flat panels, any item that has a flat hard surface
Press fit tips, adapters and vacuum hoses
Vacuum pickup hand-piece, detachable 9.6V NiMH rechargeable battery pack, In stand charger, rear exhaust filter, power on indicator, push button with on/off and lock to on, intake air filter, 8 in. wafer tip
Vacuum Tool Kit