Excelta™ Laser Line Medium Head Cutters

Small head shape allows better access while cutting in tight spaces

Manufacturer: Excelta CorporationSupplier Diversity Partner 9246E

 View more versions of this product

Catalog No. 17-467-476

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 0.01 to 0.04 in.
    Cut Type Semi flush
    Grade Extreme precision
    Grip Type ESD safe soft Foam cushion
    Head Style Medium tapered/relieved
    Includes Induction hardened to 64/65 rockwell "C" cutting blades, handles, Stainless Steel leaf springs
    Length (English) 5 in.
    Length (Metric) 12.7cm
    Material 1% Carbon tool Steel

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit