Learn More
Excelta™ Lexar Tungsten Carbide Insert Scissors
Gold plated handles for easy identification
Manufacturer:  Excelta CorporationSupplier Diversity Partner 371A
 View more versions of this product
Catalog No. 17-467-500
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Straight
15.24cm
Cutting tough substances
6 in.
Insert scissors
Stainless-steel construction with tungsten-carbide inserts