Learn More
Excelta™ MP Mirror Polished Forceps
High-quality noncorrosive stainless steel and antimagnetic
Manufacturer:  Excelta CorporationSupplier Diversity Partner 2ASAMP
 View more versions of this product
Catalog No. 17-467-124
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Straight Tapered Duckbilled Flat Tip with Round Point
Stainless Steel
Mirror-polished Forceps
0.015 in.
Extreme
0.008 in.
0.02cm
4.75 in.
12cm
Each
0.038cm
0.32 in.
0.812cm
0.06 in.