Learn More
Excelta™ Plastic Tweezers
For general precision handling without metal contact
Manufacturer:  Excelta CorporationSupplier Diversity Partner 61
 View more versions of this product
Catalog No. 16-050-115
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Flat Broad Wide Point Tip
Glass Filled Acetal
Delrin Tweezers
0.09 in.
General
1.09cm
4.62 in.
11.7cm
Each
0.22cm
0.43 in.