Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Excelta™ Precision Tweezers with Tapered Tips with High Precision Points
Precision Tweezers with Tapered Tips with High Precision Points offer extreme precision for work under microscopes.
Supplier: Excelta™ 5SSE
Description
- Antimagnetic stainless steel, Stainless steel or Carbon steel
- Straight Tapered ultra-fine tip enhances visibility under microscopes
- Overall length: 4.25 in.
Specifications
Straight Tapered, Ultra Fine | |
4.25 in. | |
108 mm | |
1 Ea. | |
0.038 cm | |
0.32 in. | |
High-precision |
5 | |
Stainless Steel | |
Tweezers, Electronic | |
0.015 in. | |
General | |
0.812 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction