Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Excelta™ Precision Tweezers with Very Fine Points
Precision Tweezers with Very Fine Points for general assembly or light magnification.
Supplier: Excelta™ 1PI
Description
- Very fine points for work under magnification or handling very fine wires and components
- Antimagnetic stainless steel, Magnetic stainless steel or Carbon Steel
- Overall length: 4.25 to 4.75 in.
Specifications
Slender, Very Fine, Straight, Precision | |
4.5 in. | |
114 mm | |
1 Ea. | |
0.03 cm | |
0.06 in. | |
0.0078 in. | |
0.019 cm | |
0.119 mm |
1 | |
Carbon Steel | |
Tweezers, Electronic | |
0.015 in. | |
Fine | |
0.32 in. | |
0.8 cm | |
0.0047 in. | |
High-precision |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction