Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Precision Tweezers with Very Fine Points

For general assembly or light magnification

Manufacturer:  Excelta CorporationSupplier Diversity Partner 3S

 View more versions of this product

Catalog No. 18-100-819


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Very Fine Tip with Precision Point
4.75 in.
12.06cm
Each
0.03cm
0.06 in.
0.0066 in.
0.016cm
3
Stainless Steel
Electronic Tweezers
0.015 in.
High
0.32 in.
0.8cm
0.0039 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit