Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Precision Tweezers with Very Fine Points

Precision Tweezers with Very Fine Points for general assembly or light magnification.

Supplier:  Excelta™ 3SA

 View more versions of this product

Catalog No. 17-456-055


Add to Cart

Description

Description

  • Very fine points for work under magnification or handling very fine wires and components
  • Antimagnetic stainless steel, Magnetic stainless steel or Carbon Steel
  • Overall length: 4.25 to 4.75 in.
Specifications

Specifications

Very Fine, Precision
4.75 in.
120 mm
1 Ea.
0.03 cm
0.06 in.
0.0066 in.
0.016 cm
0.1 mm
3
Anti-magnetic Stainless Steel
Tweezers, Electronic
0.015 in.
High
0.32 in.
0.8 cm
0.0039 in.
High-precision
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit