Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Excelta™ Precision Tweezers with Very Fine Points
Precision Tweezers with Very Fine Points for general assembly or light magnification.
Supplier: Excelta™ 3SPI
Description
- Very fine points for work under magnification or handling very fine wires and components
- Antimagnetic stainless steel, Magnetic stainless steel or Carbon Steel
- Overall length: 4.25 to 4.75 in.
Specifications
Very Fine, Precision | |
4.75 in. | |
120 mm | |
1 Ea. | |
0.03 cm | |
0.06 in. | |
0.0066 in. | |
0.016 cm | |
0.1 mm |
3 | |
Stainless Steel | |
Tweezers, Electronic | |
0.015 in. | |
Fine | |
0.32 in. | |
0.8 cm | |
0.0039 in. | |
High-precision |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction